missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SIT1 Protein

A cDNA sequence encoding the SIT1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13511-50ug

 View more versions of this product

Product Code. 15933809

  • £1320.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

SIT1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MHLSQWTRGR SRSHPGQGRS GESVEEVPLY GNLHYLQTGR LSQDPEPDQQ DPTLGGPARA AEEVMCYTSL QLRPPQGRIP GPGTPVKYSE VVLDSEPKSQ ASGPEPELYA SVCAQTRRAR ASFPDQAYAN SQPAAS
Greater than 90.0% as determined by SDS-PAGE.
50 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
SIT1 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 20% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers