missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SIT1 Protein
A cDNA sequence encoding the SIT1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13511-5ug
This item is not returnable.
View return policy
Specifications
SIT1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MHLSQWTRGR SRSHPGQGRS GESVEEVPLY GNLHYLQTGR LSQDPEPDQQ DPTLGGPARA AEEVMCYTSL QLRPPQGRIP GPGTPVKYSE VVLDSEPKSQ ASGPEPELYA SVCAQTRRAR ASFPDQAYAN SQPAAS | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SIT1 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 20% glycerol. |