missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Small muscular Protein
A DNA sequence encoding the Homo sapiens (Human) Small muscular protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8209-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_055147.1 | |
Small muscular Protein | |
Research Use Only | |
SMPX | |
Recombinant Protein | |
E. coli | |
MNMSKQPVSN VRAIQANINI PMGAFRPGAG QPPRRKECTP EVEEGVPPTS DEEKKPIPGA KKLPGPAVNL SEIQNIKSEL KYVPKAEQ | |
Greater than 80% as determined by SDS-PAGE. |
23676 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |