missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SNRPE Protein
A cDNA sequence encoding the SNRPE was constructed and used to recombinantly synthesize the protein.
£99.15 - £1320.00
Specifications
Name | SNRPE Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15943939
|
enQuireBio™
QP13549-1ug |
1 μg |
£99.15
1µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15963939
|
enQuireBio™
QP13549-5ug |
5 μg |
£155.00
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15953939
|
enQuireBio™
QP13549-50ug |
50 μg |
£1320.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
SNRPE Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SNRPE solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 5mM DTT, 1mM EDTA and 30% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAYRGQGQKV QKVMVQPINL IFRYLQNRSR IQVWLYEQVN MRIEGCIIGF DEYMNLVLDD AEEIHSKTKS RKQLGRIMLK GDNITLLQSV SN | |
Greater than 95.0% as determined by SDS-PAGE. |