missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Spectrin alpha chain, non-erythrocytic 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Spectrin alpha chain, non-erythrocytic 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6725-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_001123910.1 | |
Spectrin alpha chain, non-erythrocytic 1 Protein | |
Research Use Only | |
SPTAN1 | |
Recombinant Protein | |
E. coli | |
MDPSGVKVLE TAEDIQERRQ QVLDRYHRFK ELSTLRRQKL EDSYRFQFFQ RDAEELEKWI QEKLQIASDE NYKDPTNLQG KLQKHQAFEA EVQANSGAIV KLDETGNLMI SEGHFASETI RTRLMELHRQ WELLLEKMRE KGIKLLQAQK LVQYLRECED VMDWINDKEA IVTSEELGQD LEHVEVLQKK FEEFQTDMAA HEERVNEVNQ FAAKLIQEQH PEEELIKTKQ DEVNAAWQRL KGLALQRQGK LFGAAEVQRF NRDVDETISW IKEKEQLMAS DDFGRDLASV QALLRKHEGL ERDLAALEDK VKALCAEADR LQQSHPLSAT QIQVKREELI TNWEQIRTLA AERHARLNDS YRLQRFLADF RDLTSWVTEM KALINADELA SDVAGAEALL DRHQEHKGEI DAHEDSFKSA DESGQALLAA GHYASDEVRE KLTVLSEERA ALLELWELRR QQYEQCMDLQ LFYRDTEQVD NWMSKQEAFL LNEDLGDSLD SVEALLKKHE DFEKSLSAQE EKITALDEFA TKLIQNNHYA MEDVATRRDA LLSRRNALHE RAMRRRAQLA DSFHLQQFFR | |
Greater than 90% as determined by SDS-PAGE. |
6709 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |