missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Sphingosine Kinase 2/SPHK2 Protein

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 5298-SK-010

233.26 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18107880

  • £305.00 / 10µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Recombinant Human Sphingosine Kinase 2/SPHK2 Protein is derived from Sf 21 (baculovirus).
Specifications

Specifications

Supplied as a 0.2μm filtered solution in MES, NaCl, Glycerol and DTT.
70 kDa
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.
Recombinant
56848
Sphingosine Kinase 2/SPHK2
Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Sphingosine Kinase 2/SPHK2 protein Asn2-Pro654,with an N-terminal 5-His tag
Unconjugated
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers