missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SRGN Protein
A cDNA sequence encoding the SRGN was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13590-HIS-10ug
This item is not returnable.
View return policy
Specifications
SRGN Protein | |
Research Use Only | |
Recombinant Protein | |
Human (HEK293) | |
YPTRRARYQW VRCNPDSNSA NCLEEKGPMF ELLPGESNKI PRLRTDLFPK TRIQDLNRIF PLSEDYSGSG FGSGSGSGSG SGSGFLTEME QDYQLVDESD AFHDNLRSLD RNLPSDSQDL GQHGLEEDFM L HHHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SRGN filtered (0.4microm) solution at a concentration of 0.25 mg/ml in phosphate buffered saline and 20% (w/v) glycerol. |