missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SRI / Sorcin Protein
A DNA sequence encoding the Homo sapiens (Human) SRI / Sorcin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6727-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001243820.1 | |
SRI / Sorcin Protein | |
Research Use Only | |
SRI | |
Recombinant Protein | |
E. coli | |
MAYPGHPGAG GGYYPGGYGG APGGPAFPGQ TQDPLYGYFA AVAGQDGQID ADELQRCLTQ SGIAGGYKPF NLETCRLMVS MLDRDMSGTM GFNEFKELWA VLNGWRQHFI SFDTDRSGTV DPQELQKALT TMGFRLSPQA VNSIAKRYST NGKITFDDYI ACCVKLRALT DSFRRRDTAQ QGVVNFPYDD FIQCVMSV | |
Greater than 80% as determined by SDS-PAGE. |
6717 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |