missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human STEAP4 Protein
A cDNA sequence encoding the STEAP4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13614-20ug
This item is not returnable.
View return policy
Specifications
STEAP4 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMEKT CIDALPLTMN SSEKQETVCI FGTGDFGRSL GLKMLQCGYS VVFGSRNPQK TTLLPSGAEV LSYSEAAKKS GIIIIAIHRE HYDFLTELTE VLNGKILVDI SNNLKINQYP ESNAEYLAHL VPGAHVVKAF NTISAWALQS GALDASRQ | |
Greater than 90.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
STEAP4 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA and 10% glycerol. |