missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human STK17A Protein
A cDNA sequence encoding the STK17A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13617-2ug
This item is not returnable.
View return policy
Specifications
STK17A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSGRELGRG KFAVVRKCIK KDSGKEFAAK FMRKRRKGQD CRMEIIHEIA VLELAQDNPW VINLHEVYET ASEMILVLEY AAGGEIFDQC VADREEAFKE KDVQRLMRQI LEGVHFLHTR DVVHLDLKPQ NILLTSESPL GDIKIVDFGL SRILKNSEEL REIMGTPEYV APEILSYDPI SMATDMWSIG VLTYVMLTGI SPFLGNDKQE TFLNISQMNL SYSEEEFDVL SESAVDFIRT LLVKKPEDRA TAEECLKHPW L | |
Greater than 80.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
STK17A protein solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |