missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human STMN2 Protein
A cDNA sequence encoding the STMN2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13621-2ug
This item is not returnable.
View return policy
Specifications
STMN2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MDMEVKQINK RASGQAFELI LKPPSPISEA PRTLASPKKK DLSLEEIQKK LEAAEERRKS QEAQVLKQLA EKREHEREVL QKALEENNNF SKMAEEKLIL KMEQIKENRE ANLAAIIERL QEKERHAAEV RRNKELQVEL SG | |
Greater than 90.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The STMN2 protein solution contains 50mM Tris-HCl, pH 7.5 and 10% Glycerol. |