Learn More
R&D Systems™ Recombinant Human Survivin Western Blot Standard Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Western Blot
Brand:  R&D Systems™ WBC010B
105.40 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18196580
  • £134.00 / 100µL
Explore more special offers
Description
Description
The Recombinant Human Survivin Western Blot Standard Protein has been validated for the following applications: Western Blot.
Specifications
Specifications
332
1 Vial
API4, apoptosis inhibitor 4, Apoptosis inhibitor survivin, baculoviral IAP repeat-containing 5, Baculoviral IAP Repeat-Containing Protein 5, BIRC5, EPR-1, IAP4, survivin variant 3 alpha
Recombinant
Survivin Western Blot Standard
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.