missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SWI / SNF complex subunit SMARCC1 Protein
A DNA sequence encoding the Homo sapiens (Human) SWI/SNF complex subunit SMARCC1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7840-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003065.3 | |
SWI / SNF complex subunit SMARCC1 Protein | |
Research Use Only | |
SMARCC1 | |
Recombinant Protein | |
E. coli | |
IIIPSYASWF DYNCIHVIER RALPEFFNGK NKSKTPEIYL AYRNFMIDTY RLNPQEYLTS TACRRNLTGD VCAVMRVHAF LEQWGLVNYQ VDPESRPMAM GPPPTPHFNV LADTPSGLVP LHLRSPQVPA AQQMLNFPEK NKEKPVDLQN FGLRTDIYSK KTLAKSKGAS AGREWTEQET LLLLEALEMY KDDWNKVSEH VGSRTQDECI LHFLRLPIED P | |
Greater than 90% as determined by SDS-PAGE. |
6599 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |