missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Syntaxin-10 Protein
A DNA sequence encoding the Homo sapiens (Human) Syntaxin-10, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6741-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001258540.1 | |
Syntaxin-10 Protein | |
Research Use Only | |
STX10 | |
Recombinant Protein | |
E. coli | |
SLEDPFFVVR GEVQKAVNTA RGLYQRWCEL LQESAAVGRE ELDWTTNELR NGLRSIEWDL EDLEETIGIV EANPGKFKLP AGDLQERKVF VERMREAVQE MKDHMVSPTA VAFLERNNRE ILAGKPAAQK SPSDLLDASA VSATSRYIEE QQATQQLIMD EQDQQLEMVS GSIQVLKHMS GRVGEELDEQ GIMLDAFAQE MDHTQSRMDG VLRKLAKVSH MTSDRRQ | |
Greater than 90% as determined by SDS-PAGE. |
8677 | |
100 μg | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |