missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Syntaxin 1A Protein

Highly purified. Generating reliable and reproducible results.

£313.00 - £864.00

Specifications

Concentration 1mg/mL
For Use With (Application) SDS-PAGE
Formulation Phosphate buffered saline (pH 7.4) containing 10% glycerol, 1mM DTT
Gene ID (Entrez) 6804
Molecular Weight (g/mol) TMW: 30.7kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18052986
View Documents
Novus Biologicals™
NBP2-52366-0.02MG
0.02 mg
£313.00
0.02mg
Please to purchase this item. Need a web account? Register with us today!
18266880
View Documents
Novus Biologicals™
NBP2-52366-0.1MG
0.1 mg
£864.00
0.10mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

An un-tagged recombinant protein corresponding to the amino acids 1-265 of Human Syntaxin 1A The Recombinant Human Syntaxin 1A Protein is derived from E. coli. The Recombinant Human Syntaxin 1A Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

1mg/mL
Phosphate buffered saline (pH 7.4) containing 10% glycerol, 1mM DTT
TMW: 30.7kDa
HPC-1, Neuron-specific antigen HPC-1, p35-1, STX1P35-1, SYN1A, syntaxin 1A (brain), syntaxin-1A
Unconjugated
>95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
SDS-PAGE
6804
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Syntaxin 1A
Recombinant
Videos
SDS
Documents

Documents

Product Certifications