missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TBC1D1 Protein
A cDNA sequence encoding the TBC1D1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13692-5ug
This item is not returnable.
View return policy
Specifications
TBC1D1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSEEEAF KMLKFLMFDM GLRKQYRPDM IILQIQMYQL SRLLHDYHRD LYNHLEEHEI GPSLYAAPWF LTMFASQFPL GFVARVFDMI FLQGTEVIFK VALSLLGSHK PLILQHENLE TIVDFIKSTL PNLGLVQMEK TINQVFEMDI AKQLQAYEVE YHVLQEELID SSPLSDNQRM DKLEKTNSSL RKQNLDLLEQ LQVANGRIQS LEATIEKLLS SESKLKQAML TLELERSALL QTVEELRRRS AEPSDREPEC TQPEPTGD | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
TBC1D1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol. |