missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Telethonin Protein
A DNA sequence encoding the Homo sapiens (Human) Telethonin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6767-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003664.1 | |
Telethonin Protein | |
Research Use Only | |
TCAP | |
Recombinant Protein | |
E. coli | |
MATSELSCEV SEENCERREA FWAEWKDLTL STRPEEGCSL HEEDTQRHET YHQQGQCQVL VQRSPWLMMR MGILGRGLQE YQLPYQRVLP LPIFTPAKMG ATKEEREDTP IQLQELLALE TALGGQCVDR QEVAEITKQL PPVVPVSKPG ALRRSLSRSM SQEAQRG | |
Greater than 80% as determined by SDS-PAGE. |
8557 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |