missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TGFBI (135 Amino Acids Protein
A cDNA sequence encoding the TGFBI (135 Amino Acids was constructed and used to recombinantly synthesize the protein.
£99.15 - £3085.00
Specifications
Name | TGFBI (135 Amino Acids Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15944529
|
enQuireBio™
QP13733-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15934529
|
enQuireBio™
QP13733-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15954529
|
enQuireBio™
QP13733-EC-1MG |
1 mg |
£3085.00
1mg |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
TGFBI (135 Amino Acids Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The TGFBI recombinant Human is formulated 20mM Tris-HCl pH 8. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGTVMDVLKG DNRFSMLVAA IQSAGLTETL NREGVYTVFA PTNEAFRALP PRERSRLLGD AKELANILKY HIGDEILVSG GIGALVRLKS LQGDKLEVSL KNNVVSVNKE PVAEPDIMAT NGVVHVITNV LQPPA | |
Greater than 95.0% as determined by SDS-PAGE. |