missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Thioredoxin-related transmembrane protein 1
A DNA sequence encoding the Homo sapiens (Human) Thioredoxin-related transmembrane protein 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6810-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001137484.1 | |
Thioredoxin-related transmembrane protein 1 | |
Research Use Only | |
TMX2 | |
Recombinant Protein | |
E. coli | |
LPTQREDGNP CDFDWREVEI LMFLSAIVMM KNRRSITVEQ HIGNIFMFSK VANT | |
Greater than 90% as determined by SDS-PAGE. |
51075 | |
500 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |