missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Thioredoxin / TXN / SASP Protein
A DNA sequence encoding the Homo sapiens (Human) Thioredoxin / TXN / SASP Protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8427-ec-50ug
This item is not returnable.
View return policy
Specifications
NP_001231867.1 | |
Thioredoxin / TXN / SASP Protein | |
Research Use Only | |
TXN | |
Recombinant Protein | |
E. coli | |
VKQIESKTAF QEALDAAGDK LVVVDFSATW CGPCKMIKPF FHSLSEKYSN VIFLEVDVDD CQDVASECEV KCMPTFQFFK KGQKVGEFSG ANKEKLEATI NELV | |
Greater than 90% as determined by SDS-PAGE. |
7295 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |