missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Thyroid hormone receptor beta Protein
A DNA sequence encoding the Homo sapiens (Human) Thyroid hormone receptor beta, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6788-ec-50ug
This item is not returnable.
View return policy
Specifications
NP_000452.2 | |
Thyroid hormone receptor beta Protein | |
Research Use Only | |
THRB | |
Recombinant Protein | |
E. coli | |
MTPNSMTENG LTAWDKPKHC PDREHDWKLV GMSEACLHRK SHSERRSTLK NEQSSPHLIQ TTWTSSIFHL DHDDVNDQSV SSAQTFQTEE KKCKGYIPSY LDKDELCVVC GDKATGYHYR CITCEGCKGF FRRTIQKNLH PSYSCKYEGK CVIDKVTRNQ CQECRFKKCI YVGMATDLVL DDSKRLAKRK LIEENREKRR REELQKSIGH KPEPTDEEWE LIKTVTEAHV ATNAQGSHWK QKRKFLPEDI GQAPIVNAPE GGKVDLEAFS HFTKIITPAI TRVVDFAKKL PMFCELPCED QIILLKGCCM EIMSLRAAVR YDPESETLTL NGEMAVTRGQ LKNGGLGVVS DAIFDLGMSL SSFNLDDTEV ALLQAVLLMS SDRPGLACVE RIEKYQDSFL LAFEHYINYR KHHVTHFWPK LLMKVTDLRM IGACHASRFL HMKVECPTEL FPPLFLEVFE D | |
Greater than 90% as determined by SDS-PAGE. |
7068 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |