missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TNFRSF14 Protein
A cDNA sequence encoding the TNFRSF14 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13768-25ug
This item is not returnable.
View return policy
Specifications
8740 | |
25 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSLPSCKED EYPVGSECCP KCSPGYRVKE ACGELTGTVC EPCPPGTYIA HLNGLSKCLQ CQMCDPAMGL RASRNCSRTE NAVCGCSPGH FCIVQDGDHC AACRAYATSS PGQRVQKGGT ESQDTLCQNC PPGTFSPNGT LEECQHQTKC SWLVTKAGAG TSSSHWV | |
Greater than 85% as determined by SDS-PAGE. |
TNFRSF14 Protein | |
Research Use Only | |
TNFSF14 | |
Human | |
His | |
The TNFRSF14 solution (0.5 mg/ml) containing 20mM Tris-HCl (pH 8.0), 0.1M NaCl and 10% glycerol. |