missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Tripartite motif-containing protein 1

A DNA sequence encoding the Homo sapiens (Human) Tripartite motif-containing protein 5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8134-ec-100ug

 View more versions of this product

Product Code. 16002841

  • £384.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_149023.2
Tripartite motif-containing protein 1
Research Use Only
TRIM5
Recombinant Protein
E. coli
MASGILVNVK EEVTCPICLE LLTQPLSLDC GHSFCQACLT ANHKKSMLDK GESSCPVCRI SYQPENIRPN RHVANIVEKL REVKLSPEGQ KVDHCARHGE KLLLFCQEDG KVICWLCERS QEHRGHHTFL TEEVAREYQV KLQAALEMLR QKQQEAEELE ADIREEKASW KTQIQYDKTN VLADFEQLRD ILDWEESNEL QNLEKEEEDI LKSLTNSETE MVQQTQSLRE LISDLEHRLQ GSVMELLQGV DGVIKRTENV TLKKPETFPK NQRRVFRAPD LKGMLEVFRE LTDVRRYWVD VTVAPNNISC AVISEDKRQV SSPKPQIIYG ARGTRYQTFV NFNYCTGILG SQSITSGKHY WEVDVSKKTA WILGVCAGFQ PDAMCNIEKN ENYQPKYGYW VIGLEEGVKC SAFQDSSFHT PSVPFIVPLS VIICPDRVGV FLDYEACTVS FFNITNHGFL IYKFSHCSFS QPVFPYLNPR KCGVPMTLCS PSS
Greater than 85% as determined by SDS-PAGE.
85363
100 μg
Not determined.
Full Length
Human
His-B2M
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers