missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tropomodulin-3 Protein
A DNA sequence encoding the Homo sapiens (Human) Tropomodulin-3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8124-ec-500ug
This item is not returnable.
View return policy
Specifications
NP_055362.1 | |
Tropomodulin-3 Protein | |
Research Use Only | |
TMOD3 | |
Recombinant Protein | |
E. coli | |
MALPFRKDLE KYKDLDEDEL LGNLSETELK QLETVLDDLD PENALLPAGF RQKNQTSKST TGPFDREHLL SYLEKEALEH KDREDYVPYT GEKKGKIFIP KQKPVQTFTE EKVSLDPELE EALTSASDTE LCDLAAILGM HNLITNTKFC NIMGSSNGVD QEHFSNVVKG EKILPVFDEP PNPTNVEESL KRTKENDAHL VEVNLNNIKN IPIPTLKDFA KALETNTHVK CFSLAATRSN DPVATAFAEM LKVNKTLKSL NVESNFITGV GILALIDALR DNETLAELKI DNQRQQLGTA VELEMAKMLE ENTNILKFGY QFTQQGPRTR AANAITKNND LVRKRRVEGD HQ | |
Greater than 80% as determined by SDS-PAGE. |
29766 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |