missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tropomodulin-3 Protein
A DNA sequence encoding the Homo sapiens (Human) Tropomodulin-3, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_055362.1 |
---|---|
Gene ID (Entrez) | 29766 |
Name | Tropomodulin-3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16092791
|
enQuireBio™
QP8124-ec-10ug |
10 μg | |||||||
16032801
|
enQuireBio™
QP8124-ec-50ug |
50 μg | |||||||
16082791
|
enQuireBio™
QP8124-ec-100ug |
100 μg |
£384.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16012801
|
enQuireBio™
QP8124-ec-200ug |
200 μg | |||||||
16022801
|
enQuireBio™
QP8124-ec-500ug |
500 μg | |||||||
16002801
|
enQuireBio™
QP8124-ec-1mg |
1 mg |
£1735.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_055362.1 | |
Tropomodulin-3 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
29766 | |
Research Use Only | |
TMOD3 | |
Recombinant Protein | |
E. coli | |
MALPFRKDLE KYKDLDEDEL LGNLSETELK QLETVLDDLD PENALLPAGF RQKNQTSKST TGPFDREHLL SYLEKEALEH KDREDYVPYT GEKKGKIFIP KQKPVQTFTE EKVSLDPELE EALTSASDTE LCDLAAILGM HNLITNTKFC NIMGSSNGVD QEHFSNVVKG EKILPVFDEP PNPTNVEESL KRTKENDAHL VEVNLNNIKN IPIPTLKDFA KALETNTHVK CFSLAATRSN DPVATAFAEM LKVNKTLKSL NVESNFITGV GILALIDALR DNETLAELKI DNQRQQLGTA VELEMAKMLE ENTNILKFGY QFTQQGPRTR AANAITKNND LVRKRRVEGD HQ | |
Greater than 80% as determined by SDS-PAGE. |