missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Trypsin 3/PRSS3 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand: R&D Systems™ 3710-SE-010
This item is not returnable.
View return policy
Description
Measured by its ability to cleave the fluorogenic peptide substrate, Mca-RPKPVE-Nval-WRK(Dnp)-NH2 (Catalog # ES002 ). The specific activity is >4,000 pmol/min/μg, as measured under the described conditions.Specifications
NP_002762 | |
Supplied as a 0.2 μm filtered solution in MES and NaCl. | |
M.W. (Observed): 32 kDa, reducing conditions; M.W. (theoretical): 27 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. | |
<1.0 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
Enzyme Activity | |
5646 | |
Trypsin 3/PRSS3 | |
Mouse myeloma cell line, NS0-derived human Trypsin 3/PRSS3 protein Val16-Ser247, with a C-terminal 10-His tag | |
Pro form | |
PRSS3 | |
Recombinant | |
Trypsin 3/PRSS3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction