missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tryptophan 2,3-dioxygenase Protein
A DNA sequence encoding the Homo sapiens (Human) Tryptophan 2,3-dioxygenase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6770-ye-500ug
This item is not returnable.
View return policy
Specifications
NP_005642.1 | |
Tryptophan 2,3-dioxygenase Protein | |
Research Use Only | |
Tdo2 | |
Recombinant Protein | |
Yeast | |
MSGCPFLGNN FGYTFKKLPV EGSEEDKSQT GVNRASKGGL IYGNYLHLEK VLNAQELQSE TKGNKIHDEH LFIITHQAYE LWFKQILWEL DSVREIFQNG HVRDERNMLK VVSRMHRVSV ILKLLVQQFS ILETMTALDF NDFREYLSPA SGFQSLQFRL LENKIGVLQN MRVPYNRRHY RDNFKGEENE LLLKSEQEKT LLELVEAWLE RTPGLEPHGF NFWGKLEKNI TRGLEEEFIR IQAKEESEEK EEQVAEFQKQ KEVLLSLFDE KRHEHLLSKG ERRLSYRALQ GALMIYFYRE EPRFQVPFQL LTSLMDIDSL MTKWRYNHVC MVHRMLGSKA GTGGSSGYHY LRSTVSDRYK VFVDLFNLST YLIPRHWIPK MNPTIHKFLY TAEYCDSSYF SSDESD | |
Greater than 90% as determined by SDS-PAGE. |
6999 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |