missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TSSK6 Protein
A cDNA sequence encoding the TSSK6 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13833-5ug
This item is not returnable.
View return policy
Specifications
TSSK6 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSGDKLL SELGYKLGRT IGEGSYSKVK VATSKKYKGT VAIKVVDRRR APPDFVNKFL PRELSILRGV RHPHIVHVFE FIEVCNGKLY IVMEAAATDL LQAVQRNGRI PGVQARDLFA QIAGAVRYLH DHHLVHRDLK CENVLLSPDE RRVKLTDFGF GRQAHGYPDL STTYCGSAAY ASPEVLLGIP YDPKKYDVWS MGVVLYVMVT GCMPFDDSDI AGLPRRQKRG VLYPEGLELS ERCKALIAEL LQFSPSARPS AGQVARNCWL RAGDSG | |
Greater than 80.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The TSSK6 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |