missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human UBE2D4 Protein

A DNA sequence encoding the Homo sapiens (Human) UBE2D4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8267-ec-10ug

 View more versions of this product

Product Code. 16003661

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_057067.1
UBE2D4 Protein
Research Use Only
UBE2D4
Recombinant Protein
E. coli
MALKRIQKEL TDLQRDPPAQ CSAGPVGDDL FHWQATIMGP NDSPYQGGVF FLTIHFPTDY PFKPPKVAFT TKIYHPNINS NGSICLDILR SQWSPALTVS KVLLSICSLL CDPNPDDPLV PEIAHTYKAD REKYNRLARE WTQKYAM
Greater than 80% as determined by SDS-PAGE.
51619
10 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers