missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Ubiquitin N-Terminal Biotin Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand: R&D Systems™ UB-560-050
194.38 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Human Ubiquitin N-Terminal Biotin Protein is derived from E. coli. The Recombinant Human Ubiquitin N-Terminal Biotin Protein has been validated for the following applications: Enzyme Activity.Specifications
P0CG47.1 | |
7314 | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. | |
Recombinant |
Lyophilized from a solution in HEPES. | |
Ubiquitin N-Terminal Biotin | |
E. coli-derived human Ubiquitin protein Gln2 - Gly76 with a single N-terminal biotin | |
Biotin | |
>95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain. |