missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Ubiquitin N-Terminal Fluorescein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ U-580-050

166.74 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18370318

  • £219.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Recombinant Human Ubiquitin N-Terminal Fluorescein is derived from E. coli. The Recombinant Human Ubiquitin N-Terminal Fluorescein has been validated for the following applications: Enzyme Activity.
Specifications

Specifications

P0CG47.1
7314
Ubiquitin N-Terminal Fluorescein
E. coli-derived human Ubiquitin protein Gln2 - Gly76 with a single N-terminal fluorescein
Fluorescein
Reconstitute in aqueous buffer or DMSO to 1mg/mL
Lyophilized from a solution in deionized water.
M.W. Theroretical: 9 kDa
50 μg
Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Recombinant
95%, by PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers