missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human UFC1 Protein

A cDNA sequence encoding the UFC1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13881-20ug

 View more versions of this product

Product Code. 15954999

  • £155.00 / 20µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

UFC1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MADEATRRVV SEIPVLKTNA GPRDRELWVQ RLKEEYQSLI RYVENNKNAD NDWFRLESNK EGTRWFGKCW YIHDLLKYEF DIEFDIPITY PTTAPEIAVP ELDGKTAKMY RGGKICLTDH FKPLWARNVP KFGLAHLMAL GLGPWLAVEI PDLIQKGVIQ HKEKCNQ
Greater than 90% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
UFC1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers