missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human UNC119B Protein
A cDNA sequence encoding the UNC119B was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13894-1mg
This item is not returnable.
View return policy
Specifications
UNC119B Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSGSNPKAAA AASAAGPGGL VAGKEEKKKA GGGVLNRLKA RRQAPHHAAD DGVGAAVTEQ ELLALDTIRP EHVLRLSRVT ENYLCKPEDN IYSIDFTRFK IRDLETGTVL FEIAKPCVSD QEEDEEEGGG DVDISAGRFV RYQFTPAFLR LRTVGATVEF TVGDKPVSNF RMIERHYFRE HLLKNFDFDF GFCIPSSRNT CEHIYEFPQL SEDVIRLMIE NPYETRSDSF YFVDNKLIMH NKADYAYNGG Q | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The UNC119B solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 100mM NaCl, 1mM DTT and 10% glycerol. |