missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human USP15 Protein
A cDNA sequence encoding the USP15 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13911-1mg
This item is not returnable.
View return policy
Specifications
USP15 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAEGGAA DLDTQRSDIA TLLKTSLRKG DTWYLVDSRW FKQWKKYVGF DSWDKYQMGD QNVYPGPIDN SGLLKDGDAQ SLKEHLIDEL DYILLPTEGW NKLVSWYTLM EGQEPIARKV VEQGMFVKHC KVEVYLTELK LCENGNMNNV VTRRFSKADT IDTIEKEIRK IFSIPDEKET RLWNKYMSNT FEPLNKPDST IQDAGLYQGQ VLVIEQKNED GTWPRGPSTP KKPLEQSC | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The USP15 solution contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 10% glycerol. |