Learn More
R&D Systems™ Recombinant Human VCAM-1/CD106 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 809-VR-050
This item is not returnable.
View return policy
Description
The Recombinant Human VCAM-1/CD106 Protein is derived from NS0. The Recombinant Human VCAM-1/CD106 Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS containing Calcium and Magnesium with Sorbitol. | |
74 kDa | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
7412 | |
VCAM-1/CD106 | |
Mouse myeloma cell line,NS0-derived human VCAM-1/CD106 protein Phe25-Glu698,with a substitution of Arg322Gly and an N-terminal Met | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |