missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human VEGF 165 Protein
Brand: R&D Systems™ BT-VEGF-020
220.30 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Measured in a cell proliferation assay using HUVEC human umbilical vein endothelial cells. Conn, G. et al. (1990) Proc. Natl. Acad. Sci. USA 87:1323. The ED50 for this effect is 1.50-12.0 ng/mL.Specifications
NP_001165097.1 | |
Lyophilized from a 0.2 μm filtered solution in Sodium Acetate. | |
M.W. Observed: 19-21 kDa, under reducing conditions., M.W. Theoretical: 19.2 kDa (monomer) | |
E. coli-derived human VEGF protein Ala27-Arg191 | |
<0.01 EU per 1μg of the protein by the LAL method. | |
Unconjugated | |
>97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
Bioactivity | |
7422 | |
20 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor | |
Reconstitute the 10 μg size at 100 μg/mL in sterile deionized water. Reconstitute all other sizes at 500 μg/mL in sterile deionized water. | |
VEGF |