missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human VEGF 165 Protein

Brand:  R&D Systems™ BT-VEGF-100

826.76 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.


 View more versions of this product

Product Code. 18789886

  • £1050.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Measured in a cell proliferation assay using HUVEC human umbilical vein endothelial cells. Conn, G. et al. (1990) Proc. Natl. Acad. Sci. USA 87:1323. The ED50 for this effect is 1.50-12.0 ng/mL.
Specifications

Specifications

NP_001165097.1
Lyophilized from a 0.2 μm filtered solution in Sodium Acetate.
M.W. Observed: 19-21 kDa, under reducing conditions., M.W. Theoretical: 19.2 kDa (monomer)
E. coli-derived human VEGF protein Ala27-Arg191
<0.01 EU per 1 μg of the protein by the LAL method.
Unconjugated
>97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie® Blue Staining.
Bioactivity
7422
100 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
MVCD1, VAS, vascular endothelial growth factor A, Vascular permeability factor, Vasculotropin, VEGFA, VEGF-A, VEGFMGC70609, VPF, VPFvascular endothelial growth factor
Reconstitute the 10 μg size at 100 μg/mL in sterile deionized water. Reconstitute all other sizes at 500 μg/mL in sterile deionized water.
VEGF
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers