missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human VNN1 Protein
A cDNA sequence encoding the VNN1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13938-1mg
This item is not returnable.
View return policy
Specifications
VNN1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHASQDTFTAAVYE HAAILPNATL TPVSREEALA LMNRNLDILE GAITSAADQG AHIIVTPEDA IYGWNFNRDS LYPYLEDIPD PEVNWIPCNN RNRFGQTPVQ ERLSCLAKNN SIYVVANIGD KKPCDTSDPQ CPPDGRYQYN TDVVFDSQGK LVARYHKQNL FMGENQFNVP KEPEIVTFNT TFGSFGIFTC FDILFHDPAV TLVKDFHVDT IVFPTAWMNV LPHLSAVEFH SAWAMGMRVN FLASNIHYPS KKMTGSGIYA PNSSRAFHYD MKTEEGKLLL SQLDSHPSHS AVVNWTSYAS SIEALSSGNK EFKGTVFFDE FTFVKLTGVA GNYTVCQKDL CCHLSYKMSE NIPNEVYALG AFDGLHTVEG RYYLQICTLL KCKTTNLNTC GDSAETASTR FEMFSLSGTF GTQYVFPEVL LSENQLAPGE FQVSTDGRLF SLKPTSGPVL TVTLFGRLYE KDWASNASSG | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
VNN1 was filtered (0.4 microm) and lyophilized in 20mM Tris buffer, 50mM NaCl and 0.1% amisoft CS-22, pH 7.5. |