missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human WIBG Protein
A cDNA sequence encoding the WIBG was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13958-1mg
This item is not returnable.
View return policy
Specifications
WIBG Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MEAAGSPAAT ETGKYIASTQ RPDGTWRKQR RVKEGYVPQE EVPVYENKYV KFFKSKPELP PGLSPEATAP VTPSRPEGGE PGLSKTAKRN LKRKEKRRQQ QEKGEAEALS RTLDKVSLEE TAQLPSAPQG SRAAPTAASD QPDSAATTEK AKKIKNLKKK LRQVEELQQR IQAGEVSQPS KEQLEKLARR RALEEELEDL ELGLLEHHHH HH | |
Greater than 85.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
WIBG Human solution containing 20mM Tris pH 8.0, 0.1M NaCl and 10% glycerol. |