missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human WIBG Protein
A cDNA sequence encoding the WIBG was constructed and used to recombinantly synthesize the protein.
£99.15 - £5730.00
Specifications
Name | WIBG Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15965239
|
enQuireBio™
QP13958-2ug |
2 μg |
£99.15
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15945239
|
enQuireBio™
QP13958-10ug |
10 μg |
£155.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15955239
|
enQuireBio™
QP13958-1mg |
1 mg |
£5730.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
WIBG Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
WIBG Human solution containing 20mM Tris pH 8.0, 0.1M NaCl and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MEAAGSPAAT ETGKYIASTQ RPDGTWRKQR RVKEGYVPQE EVPVYENKYV KFFKSKPELP PGLSPEATAP VTPSRPEGGE PGLSKTAKRN LKRKEKRRQQ QEKGEAEALS RTLDKVSLEE TAQLPSAPQG SRAAPTAASD QPDSAATTEK AKKIKNLKKK LRQVEELQQR IQAGEVSQPS KEQLEKLARR RALEEELEDL ELGLLEHHHH HH | |
Greater than 85.0% as determined by SDS-PAGE. |