missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human WLS Protein

A cDNA sequence encoding the WLS was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13962-2ug

 View more versions of this product

Product Code. 15985249

  • £99.15 / 2µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

WLS Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMEMSPWF QFMLFILQLD IAFKLNNQIR ENAEVSMDVS LAYRDDAFAE WTEMAHERVP RKLKCTFTSP KTPEHEGRYY ECDVLPFMEI GSVAHKFYLL NIRLPVNEKK KINVGIGEIK DIRLVGIHQN GGFTK
Greater than 90% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
WLS protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0),0.4M Urea and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers