missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human WLS Protein
A cDNA sequence encoding the WLS was constructed and used to recombinantly synthesize the protein.
£99.15 - £5730.00
Specifications
Name | WLS Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15985249
|
enQuireBio™
QP13962-2ug |
2 μg |
£99.15
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15965249
|
enQuireBio™
QP13962-10ug |
10 μg |
£155.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15975249
|
enQuireBio™
QP13962-1mg |
1 mg |
£5730.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
WLS Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
WLS protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0),0.4M Urea and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMEMSPWF QFMLFILQLD IAFKLNNQIR ENAEVSMDVS LAYRDDAFAE WTEMAHERVP RKLKCTFTSP KTPEHEGRYY ECDVLPFMEI GSVAHKFYLL NIRLPVNEKK KINVGIGEIK DIRLVGIHQN GGFTK | |
Greater than 90% as determined by SDS-PAGE. |