missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Wnt-3a Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£296.00 - £4335.00
Specifications
Gene ID (Entrez) | 89780 |
---|---|
Molecular Weight (g/mol) | 37.4 kDa |
Name | Wnt-3a |
Source | Chinese Hamster Ovary cell line,CHO-derived human Wnt-3a protein Ser19-Lys352 |
Conjugate | Unconjugated |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18166560
|
R&D Systems™
5036-WN-010 |
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS with BSA as a carrier protein. |
£296.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18187340
|
R&D Systems™
5036-WN-010/CF |
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS. |
£296.00
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18694739
|
R&D Systems™
5036-WN-500/CF |
Lyophilized from a 0.2 μm filtered solution in PBS, EDTA and CHAPS. |
£4335.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18755707
|
R&D Systems™
5036-WN-500 |
Lyophilized from a 0.2 μm filtered solution in PBS, EDTA and CHAPS with BSA as a carrier protein. |
£4335.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Measured by its ability to induce alkaline phosphatase production by MC3T3-E1 mouse preosteoblast cells. The ED50 for this effect is 5.00-25.0 ng/mL. Measured by its ability to induce Topflash reporter activity in HEK293T human embryonic kidney cells. The ED50 for this effect is <500 ng/mL. Protein concentrations should be titrated based on cell type and if appropriate, passage number of the cell line. Optimal concentrations should be determined by each laboratory for each application.Specifications
89780 | |
Wnt-3a | |
Unconjugated | |
75%, by SDS-PAGE with silver staining. |
37.4 kDa | |
Chinese Hamster Ovary cell line,CHO-derived human Wnt-3a protein Ser19-Lys352 | |
Recombinant |