missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human XRP2 / RP2 Protein
A DNA sequence encoding the Homo sapiens (Human) XRP2 / RP2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6617-ec-100ug
This item is not returnable.
View return policy
Specifications
NP_008846.2 | |
XRP2 / RP2 Protein | |
Research Use Only | |
RP2 | |
Recombinant Protein | |
E. coli | |
GCFFSKRRKA DKESRPENEE ERPKQYSWDQ REKVDPKDYM FSGLKDETVG RLPGTVAGQQ FLIQDCENCN IYIFDHSATV TIDDCTNCII FLGPVKGSVF FRNCRDCKCT LACQQFRVRD CRKLEVFLCC ATQPIIESSS NIKFGCFQWY YPELAFQFKD AGLSIFNNTW SNIHDFTPVS GELNWSLLPE DAVVQDYVPI PTTEELKAVR VSTEANRSIV PISRGQRQKS SDESCLVVLF AGDYTIANAR KLIDEMVGKG FFLVQTKEVS MKAEDAQRVF REKAPDFLPL LNKGPVIALE FNGDGAVEVC QLIVNEIFNG TKMFVSESKE TASGDVDSFY NFADIQMGI | |
Greater than 80% as determined by SDS-PAGE. |
6102 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |