missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human YWHAZ Protein
A cDNA sequence encoding the YWHAZ was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13987-20ug
This item is not returnable.
View return policy
Specifications
7534 | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSHMDK NELVQKAKLA EQAERYDDMA ACMKSVTEQG AELSNEERNL LSVAYKNVVG ARRSSWRVVS SIEQKTEGAE KKQQMAREYR EKIETELRDI CNDVLSLLEK FLIPNASQAE SKVFYLKMKG DYYRYLAEVA AGDDKKGIVD QSQQAYQEAF EISKKEMQPT HPIRLGLALN FSVFYYEILN SPEKACSLAK TAFDEAIAEL DTLSEESYKD STLIMQLLRD NLTLWTSDTQ GDEAEAGEGG EN | |
Greater than 95.0% as determined by SDS-PAGE. |
YWHAZ Protein | |
Research Use Only | |
YWHAZ | |
Human | |
His | |
YWHAZ solution containing 1xPBS pH 7. |