missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ZFAND1 Protein
A cDNA sequence encoding the ZFAND1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13993-10ug
This item is not returnable.
View return policy
Specifications
ZFAND1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAELDIG QHCQVEHCRQ RDFLPFVCDD CSGIFCLEHR SRESHGCPEV TVINERLKTD QHTSYPCSFK DCAERELVAV ICPYCEKNFC LRHRHQSDHE CEKLEIPKPR MAATQKLVKD IIDSKTGETA SKRWKGAKNS ETAAKVALMK LKMHADGDKS LPQTERIYFQ VFLPKGSKEK SKPMFFCHRW SIGKAIDFAA SLARLKNDNN KFTAKKLRLC HITSGEALPL DHTLETWIAK EDCPLYNGGN IILEYLNDEE QFCKNVESYL E | |
Greater than 90.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ZFAND1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT. |