missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza A H1N1 (Hickox / 1940) Hemagglutinin Protein
A DNA sequence encoding the Influenza A virus (strain A/Hickox/1940 H1N1) Influenza A H1N1 (strain A/Hickox/1940) Hemagglutinin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7521-ec-100ug
This item is not returnable.
View return policy
Specifications
Influenza A H1N1 (Hickox / 1940) Hemagglutinin Protein | |
Research Use Only | |
HA | |
Recombinant Protein | |
E. coli | |
DTICIGYHAN NSTDTVDTVL EKNVTVTHSV NLLEDSHNGK LCRLKGIAPL QLGKCNIAGW ILGNPECESL LSKRSWSYIA ETPNSENGTC YPGDFADYEE LREQLSSVSS FERFEIFPKE RSWPNHNINI GVTAACSHAG KSSFYKNLLW LTEKDGSYPN LNKSYVNKKE KEVLVLWGVH HPSNIENQKT LYRKENAYVS VVSSNYNRRF TPEIAERPKV RGQAGRMNYY WTLLEPGDTI IFEANGNLIA PWYAFALSRG LGSGIITSNA SMDECDTKCQ TPQGAINSSL PFQNIHPFTI GECPKYVRST KLRMVTGLRN IPSIQS | |
Greater than 90% as determined by SDS-PAGE. |
100 μg | |
Not determined. | |
Full Length | |
Influenza | |
His-SUMO | |
Tris-based buffer, 50% glycerol |