missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Influenza H3N2 Switzerland Recombinant Protein

A cDNA sequence encoding the H3N2 Switzerland Recombinant was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12087-2ug

 View more versions of this product

Product Code. 15949328

  • £99.15 / 2µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Influenza H3N2 Switzerland Protein
Research Use Only
Recombinant Protein
Baculovirus
The Recombinant H3N2 A/ Switzerland 2013 solution contains 10mM Sodium phosphate, pH 7.1,150mM NaCl and 0.005% Tween-20.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Influenza
Untagged
Greater than 90.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers