missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Influenza H5N1 Indonesia Protein

A cDNA sequence encoding the H5N1 Indonesia was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12093-100ug

 View more versions of this product

Product Code. 15929348

  • £1985.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Influenza H5N1 Indonesia Protein
Research Use Only
Recombinant Protein
Baculovirus
The Recombinant H5N1 A/Indonesia/05/2005 solution contains 10mM Sodium phosphate, pH 7.2, 150mM NaCl and 0.005% Tween-20.
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Influenza
Untagged
Greater than 90.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers