missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Influenza H9N2 Hong-Kong Protein
A cDNA sequence encoding the H9N2 Hong-Kong was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12097-2ug
This item is not returnable.
View return policy
Specifications
Influenza H9N2 Hong-Kong Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
The Recombinant H9N2 A/Hong-Kong/1073/99 solution contains 10mM Sodium Phosphate, pH 7.0 and 150mM NaCl and 0.01% Tween-20. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Influenza | |
Untagged | |
Greater than 90.0% as determined by SDS-PAGE. |